PAH columns, ZORBAX® Eclipse
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
959763-918 959757-318 959758-318
HEWL959763-918EA 861 EUR
HEWL959763-918 HEWL959757-318 HEWL959758-318
PAH columns, ZORBAX® Eclipse
Kolommen voor chromatografie
ZORBAX® Eclipse PAH HPLC columns are the ideal first choice for method development of all samples.
  • High level of performance peak shape, efficiency, resolution, and lifetime with all sample types: acids, bases and neutrals
  • Superior reproducibility with more rigorous QA/QC testing
  • Improved, patented silica manufacturing with start-to-finish product control
  • Three different reversed-phase chemistries are available (Eclipse Plus C18, Eclipse Plus C8 and Eclipse Plus Phenyl-Hexyl)
  • PAH column for high resolution separation of 16 PAHs in EPA 610 - specifically