GC columns, J&W CP-Wax 57 CB
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP8120 CP97658 CP97763 CP97773 CP97721
VARICP8120EA 1300 EUR
VARICP8120 VARICP97658 VARICP97763 VARICP97773 VARICP97721
GC columns, J&W CP-Wax 57 CB
Kolommen voor chromatografie
CP-Wax 57 CB has a unique selectivity, especially for the analysis of alcohols in the brewing and wines/spirits industry.
  • Unique high polarity bonded wax column
  • Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
The high inertness of this column offers excellent peak shapes for these compounds, ensuring high precision. The 0.15 mm I.D version offers a significant gain in analysis speed.