GC columns, J&W CP-Sil 24 CB
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7821 CP7824 CP7832 CP7834 CP7871 CP7834I5 CP7831 CP7820 CP7822 CP7822I5 CP7830 CP7871I5
VARICP7821EA 737 EUR
VARICP7821 VARICP7824 VARICP7832 VARICP7834 VARICP7871 VARICP7834I5 VARICP7831 VARICP7820 VARICP7822 VARICP7822I5 VARICP7830 VARICP7871I5
GC columns, J&W CP-Sil 24 CB
Kolommen voor chromatografie
CP-Sil 24 CB is a medium polarity column with no cyano groups, making it ideal for use with ECD, and perfect for confirmation in drug and pesticide analysis.
  • Phenyl 50%/50% dimethylpolysiloxane
  • Mid polarity phase
  • Specially suitable for analysis of amines, drugs and pesticides
  • Ideal for analysis using ECD
  • Excellent confirmation column in combination with CP-Sil 5 CB or CP-Sil 8 CB
J&W CP-Sil 24 CB is a medium polarity, 50% phenyl/50% dimethylpolysiloxane phase with no cyano groups, making it ideal for use with ECD. The column produces perfect peak shapes for amines as shown by the grob test mixture. It is especially suitable for the analysis of drugs and pesticides and is an excellent confirmation column in combination with CP-Sil 5 CB or CP-Sil 8 CB.