GC columns, J&W HP-50+
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091L-113 19091L-133 19091L-333 19091L-413 19091L-413E 19091L-416 19091L-431 19091L-433 19095L-021 19095L-023 19095L-023E 19095L-523
HEWL19091L-113EA 796 EUR
HEWL19091L-113 HEWL19091L-133 HEWL19091L-333 HEWL19091L-413 HEWL19091L-413E HEWL19091L-416 HEWL19091L-431 HEWL19091L-433 HEWL19095L-021 HEWL19095L-023 HEWL19095L-023E HEWL19095L-523
GC columns, J&W HP-50+
Kolommen voor chromatografie
HP-50+ is excellent for confirmational analyses of polar compounds such as chlorinated pesticides, herbicides, and basic drugs.
  • (50%-Phenyl)-methylpolysiloxane
  • Mid-polarity - slightly less polar than DB-17
  • Excellent for confirmational analyses
  • Bonded and cross-linked
  • Solvent rinsable
J&W HP 50+ is a (50% phenyl) methylpolysiloxane phase which is excellent for confirmational analysis. These mid-polarity phase columns are generally used for analysis of polar compounds such as chlorinated pesticides, herbicides, and basic drugs. They are often used in conjunction with a second column to provide confirmational information. Additionally, the phase ratios of the 0,32 mm series are maintained in the 0,20 mm series, simplifying transfer of methods to GC/MS systems.