Capillary tubings, stainless steel, J&W UltiMetal plus
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP6577 CP6580 CP6575 CP6572 CP6578 CP6579 CP6576
VARICP6577EA 162 EUR
VARICP6577 VARICP6580 VARICP6575 HEWLCP6572 VARICP6578 VARICP6579 VARICP6576
Capillary tubings, stainless steel, J&W UltiMetal plus
Slangen Chromatography Tubing
UltiMetal Plus Stainless Steel Capillary Tubing products are the preferred choice for most applications due to its inertness and robustness.
  • UltiMetal Plus deactivation
  • Chemical deactivation provides an inert surface for analysis of active analytes at trace levels
  • Virtually unbreakable stainless steel tubing which provides robustness
  • Three types of products available: transfer lines, with ¹/₁₆" outer diameter, guard columns and 50 m capillary tubing
These tubing products can be used as transfer lines, guard columns or for making connections in GC. The stainless steel tubing products are treated with UltiMetal Plus deactivation and can be used in an inert flow path.