Peristaltic pump tubing and sample line supplies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G1820-65105 G1820-65217 G1820-65478 5064-8024 5064-8023 G1820-65527 G1820-65531 G1833-65572 G1833-65573 G1820-65526 G3138-65129 G3280-60590 G3280-40103 G8400-40113
HEWLG1820-65105EA 104 EUR
HEWLG1820-65105 HEWLG1820-65217 HEWLG1820-65478 HEWL5064-8024 HEWL5064-8023 HEWLG1820-65527 HEWLG1820-65531 HEWLG1833-65572 HEWLG1833-65573 HEWLG1820-65526 HEWLG3138-65129 HEWLG3280-60590 HEWLG3280-40103 HEWLG8400-40113
Peristaltic pump tubing and sample line supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The peristaltic pump tubing has to be washed prior to use. The common way is to run about 5% HNO₃ (v/v) solution.
  • Long lifetime
  • Resistant to most chemicals
  • Not suitable for sample introduction - impurity level is high
If customers use HCl, Sn is eluted from the Tygon tube. In this case, 3% HCl (v/v) should be pumped at 0,1 rps for less than an hour by the new peristaltic pump tube.
Opgelet: Some chemicals attack the peristaltic pump tube. This might cause leakage in the sample introduction and drain lines and corrosion of the sample introduction system.