DEA plates, Bond Elut
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7541702C 7541701C
VARI7541702CEA 554 EUR
VARI7541702C VARI7541701C
DEA plates, Bond Elut
Microplaten Platen voor vastefase-extractie
Bond Elut DEA (diethylaminopropyl) can be used as both a weak anion exchange sorbent with aqueous samples or as a polar sorbent with nonpolar organic matrixes.
  • Weak anion exchanger
  • More polar than C8 but less polar than C2 or CN
  • Alkyl side chains confer moderately nonpolar characteristics
Bond Elut DEA bears some resemblance to Bond Elut NH₂ in its properties, but with a slightly lower capacity as an anion-exchange sorbent. DEA has a moderately nonpolar character due to the alkyl side chains on the amino functionality. These groups still afford aMore Product Information