Torches for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0552 8003-0553 8003-0554
HEWL8003-0552EA 370 EUR
HEWL8003-0552 HEWL8003-0553 HEWL8003-0554
Torches for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These torches are suitable for PerkinElmer ICP-MS systems.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage