High efficiency GC columns
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP5800 CP7802 CP9033 CP7687 CP740510 CP8931 CP915429 CP9231
VARICP5800EA 601 EUR
VARICP5800 VARICP7802 VARICP9033 VARICP7687 VARICP740510 VARICP8931 VARICP915429 VARICP9231
High efficiency GC columns
Kolommen voor chromatografie
High efficiency 0,15 and 0,18 mm Int.Ø columns are a simple and cost effective way to increase sample throughput with existing GC or GC/MS instruments.
  • Increase sample throughput and productivity while maintaining resolution
  • Reduce carrier gas usage and cost per analysis
  • Operates with He or H₂ carrier gas
  • Available in more than 20 stationary phases
This leading-edge column technology is ideal for applications that require faster run times, such as high-throughput screening, fast process monitoring, and fast method development. Agilent High EfficiencyMore Product Information
Order Now
Specificaties
Environmentally Preferable