AdEasy Adenoviral vector systems
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240008 240009 240010 240005 240006 240007 240008 240009 240010 240081 240082 240085
STRA240008EA 0 EUR
STRA240008 STRA240009 STRA240010 AGLS240005 AGLS240006 AGLS240007 AGLS240008 AGLS240009 AGLS240010 AGLS240081 AGLS240082 AGLS240085
AdEasy Adenoviral vector systems
Vectoren, Plasmiden en Bibliotheken
The AdEasy system saves you a month of work over traditional methods by producing the recombinant adenoviral plasmid by homologous recombination in E. coli. The AdEasy XL system includes BJ5183 cells pre-transformed with the pAdEasy-1 plasmid, this feature dramatically decreases background caused by the non-recombinant shuttle plasmid.
  • 300% more recombinants with specialty competent cells
  • Adherent AD-293 cells improve viral packaging
  • Infects dividing and non-dividing cells
Opgelet: For Research Use Only. Not for Use in Diagnostic Procedures.