VWR®, Cell culture media, Ham's F12
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
392-0418EA 10.2 EUR
392-0418
VWR®, Cell culture media, Ham's F12
Celcultuurmedia
Ham's F12 was developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells. It is frequently used with supplements for serum-free cultures to support the growth of cells of rodent origin, or as a cloning medium for the culture of myeloma and hybrid cells.