GC columns, J&W CP-1301
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP8605 CP8613 CP8607
VARICP8605EA 1310 EUR
VARICP8605 VARICP8613 VARICP8607
GC columns, J&W CP-1301
Kolommen voor chromatografie
Agilent JW CP-1301 is a medium polarity column ideal for the analysis of herbicides, pesticides, and many pharmaceutical products.
  • 6% cyanopropyl-phenyl/94% dimethylpolysiloxane
  • Good inertness for better quality of data, even with thick films
  • Bonded and cross-linked
It is uses a bonded, 6% cyanopropyl-phenyl phase to deliver lower bleed and improved column-to-column reproducibility.