Telkamer, Neubauer
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40441702 92044102702
631-0925EA 40.2 EUR
631-0925 HECH92044102702
Telkamer, Neubauer
Object- en dekglaasjes Telkamers
These counting chambers are designed for blood tests and urine analysis.
  • Counting erythrocytes corresponds exactly to Thoma method
  • Counting leucocytes in five large squares
  • Evaluation just as indicated for Thoma ruling
Consisting of nine large squares – The central square shows the Thoma ruling and the other eight squares are subdivided.