Quick-lock connectors
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
360179 360180 360183 360189 360190
SCTV360179EA 34.9 EUR
SCTV360179 SCTV360180 SCTV360183 SCTV360189 SCTV360190
Quick-lock connectors
Fittingen en connectoren
Easy and safe connection of capillary tubing and SCAT Safety Caps using quick couplings. Connect and disconnect with a "click" - without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
  • Made of chemically resistant glass-filled polypropylene, PEEK® materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the
More Product Information
Order Now
Specificaties
Environmentally Preferable