Dialysis cassettes, Slide-A-Lyzer™ G2, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87731 87728 88251
514-0205EA 212 EUR
514-0205 514-0202 PIER88251
Dialysis cassettes, Slide-A-Lyzer™ G2, Pierce™
Slangen Dialyseslangen
The Slide-A-Lyzer® G2 dialysis cassette offers efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.
  • Pipette-accessible for easy sample loading and retrieval
  • Self-floating chambers for buoyancy and vertical orientation during dialysis
  • Designed to maintain the highest possible sample integrity and protection with rigorous quality testing for maximum consistency
  • Fast and consistent dialysis with maximum sample recovery
  • Ideal for removing low-molecular weight contaminants, performing buffer exchange and desalting
The cassette membrane is composed of low-binding regenerated cellulose for maximum sample recovery while maintaining maximum sample purity and are manufactured using clean-room conditions to ensure cassettes are contaminant-free. The Slide-A-Lyzer® G2 dialysis cassettes are available in four precise membrane MWCOs (2, 3,5, 10 and 20 K) for dialysing sample volumes up to 30 ml.
Volume range using a pipette: 15 to 30 ml
Volume range using a syringe: 10 to 30 ml
Cassettes for other sample sizes are also available. Please enquire for further details.