Celite® Standard Super-cel, filter aid, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22556.290EA 34.3 EUR
22556.290 22556.360
Celite® Standard Super-cel, filter aid, GPR RECTAPUR®
Celite® standard super-cel, filter aid
Celite® is used as a filtering aid or as adsorbent in column chromatography.
  • Filter aid
  • Calcined, fresh water diatomeous earth
MDL Nummer: MFCD00132803
CAS nummer: 91053-39-3
Order Now
Specification Test Results
pH (25 °C; 10% in water ) 8,5 - 11,0
Wet density ≤ 357 g/l
Loss on ignition (1000 °C) ≤ 0.5%
Particle size (< 56µm) ≥ 90%