Thermocouple thermometer probes, H-B Durac®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B609011200 B609011300
620-2821EA 210 EUR
620-2821 620-2822
Thermocouple thermometer probes, H-B Durac®, SP Bel-Art
Temperatuursondes
For use with H-B Durac® Calibrated Thermocouple Thermometers or others that accept K or J type probes.
  • For temperature measurement of liquids, semi-solids, or air/gasses
  • Measure wide temperature ranges
  • 15,24 cm (6") stainless steel probe