Microplates, 24-well, Whatman™ UNIFILTER®
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7700-9901 7700-9904 7700-9917
734-2549EA 1070 EUR
734-2549 734-2550 734-2551
Microplates, 24-well, Whatman™ UNIFILTER®
Microplaten Filtreerplaten
The 10 ml UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
  • Polypropylene microplate is heat and chemical resistant
  • 24 well microplate with 10 ml wells appropriate for large sample volumes
  • No cross talk: patented integral filter drip director design prevents well-to-well cross contamination
  • Filtration media options include glass microfiber, melt-blown polypropylene, PTFE, VFE
Applications such asMore Product Information
Order Now
Specificaties
Environmentally Preferable