MicroPlates, 96-well, Whatman™ UNIFILTER®
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7700-2801 7700-2803 7700-2808 7700-2810 7700-2804
734-2522EA 1150 EUR
734-2522 734-2523 734-2524 734-2525 738-0200
MicroPlates, 96-well, Whatman™ UNIFILTER®
Microplaten Filtreerplaten
UNIFILTER 96-well microplate, 800 µl, long drip director.
  • No cross talk: Patented integral filter design prevents well-to-well cross contamination
  • Economical to use: Wide range of well volume options ensures efficient use of materials
  • Better control: Choice of filter media allows control of the flow rates and retention characteristics
  • Versatile: A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose
The 800 µl UNIFILTER microplate is most typically used in purifications, isolations, and separations of biomolecules, particularly DNA.
The microplate has a well volume of 800 μl, which is excellent for standard DNA plasmid miniprep chemistries. The choice of short or long drip directors is application-specific. UNIFILTER 800 μl is constructed from rigid high-grade polystyrene.
Proprietary Whatman UNIFILTER microplates with filter bottom wells are convenient and ready to use. Available in 24-, 96-, and 384-well formats, UNIFILTER microplates offer a choice of filter media to meet exact application requirements.
The novel drip director design of Whatman UNIFILTER microplates ensures precise collection of the filtrate to allow for further processing and analysis.
Whatman UNIFILTER microplates from Cytiva's business are available in other plate materials and well formats. Well volumes range from 100 μl to 10 ml.