Fine dosage syringes, Injekt® Luer Duo
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4645022V 4645057V 4645103V 4645200V
BRAU4645022VEA 9.7 EUR
BRAU4645022V BRAU4645057V BRAU4645103V BRAU4645200V
Fine dosage syringes, Injekt® Luer Duo
Spuiten Injectiespuiten, medisch
2-piece hypodermic syringes with Luer connector for single use with detached hypodermic Sterican® needle.
  • Green plunger for precise dosage
  • Highly transparent barrel with black graduation for easy reading
  • Safe plunger backstop to prevent unintentional plunger withdrawal
  • Graduation extended beyond
  • nominal volume
  • Not manufactured with silicone oil,
  • PVC, Latex and DEHP
Barrel made of PP, plunger made of PE.
Verpakking: Sterile single packaging in box of 100 units.