Accessories for Safety Funnels with Lid 'MARCO' V2.0
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
318999 318989 319000
SCTV318999EA 11.9 EUR
SCTV318999 SCTV318989 SCTV319000
Accessories for Safety Funnels with Lid 'MARCO' V2.0
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Replacement part for SCAT 'MARCO' V2.0 safety funnels, which can be closed safely and prevent danger of ignition.
  • Suitable for all models of the MARCO series
  • Available in conductive and non-conductive versions
  • Stainless steel sieve to catch small objects such as stirrers from HPLC equipment
The 'MARCO' V2.0 safety funnel (sold separately) is a lid funnel for the disposal of liquid waste. The lid seals the funnel and prevents the escape of toxic or explosive vapours. Numerous features guarantee safe handling. Available in different thread sizes forMore Product Information