Syringe filters, prefilter, non-sterile, Whatman™ Anotop Plus
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6809-3002 6809-3012 6809-3022 6809-4002 6809-4012 6809-4022 6809-4024
516-1133EA 698 EUR
516-1133 516-1134 516-1281 516-1511 516-1513 516-1515 516-1516
Syringe filters, prefilter, non-sterile, Whatman™ Anotop Plus
Filters Spuitfilters
Whatman Anotop Plus non-sterile syringe filters from Cytiva have an integrated glass microfiber prefilter and inorganic membrane, enabling filtration of a wide variety of sample solvents and compounds.
  • Capillary pore structure for well-defined particle filtration
  • Low protein binding filter for applications requiring high protein recovery
  • Integrated glass microfiber prefilter for filtering larger volumes of difficult-to-filter samples
  • Recyclable after use
High solid content samples can be difficult to filter because of their tendency to prematurelyMore Product Information