Swing samplers
Leverancier: NASCO FORT ATKINSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B01310WA B01366WA
NASCB01310WAEA 440 EUR
NASCB01310WA NASCB01366WA
Swing samplers
Monsternemers Monsternemers voor vloeistoffen
For collecting liquids, powders, or small solids from a horizontal flowing stream
The end of the sampler swings to allow collection from different angles, including 90°. Samplers come with an extending fiberglass pole and one 960mL (32.5oz.) polyethylene bottle with plastic snapper ring.