QuickSeal Foil PCR Adhesive PCR Foil Seals
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-127-080LS IST-127-080TS
ISTSIST-127-080LSEA 127 EUR
ISTSIST-127-080LS ISTSIST-127-080TS
QuickSeal Foil PCR Adhesive PCR Foil Seals
Afdichtingen Afdichtingen van microtiterplaten
Pierceable, peelable barrier foil for PCR and sample storage.
  • Seal integrity range: −80 to +120 °C
  • Available sterile or non sterile
New Product
Order Now
Specificaties
Environmentally Preferable