Tubing rolls, FEP, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S2020-24 S2020-28 S2020-36 S2020-48 S2020-56 S2020-60 S2022-04 S2022-07 S2022-08 S2022-12 S2022-20
BOHLS2020-24EA 173 EUR
BOHLS2020-24 BOHLS2020-28 BOHLS2020-36 BOHLS2020-48 BOHLS2020-56 BOHLS2020-60 BOHLS2022-04 BOHLS2022-07 BOHLS2022-08 BOHLS2022-12 BOHLS2022-20
Tubing rolls, FEP, BOLA
Slangen Transfer Tubing
Non-porous, transparent, gastight tubing.
  • Most universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Non-adhesive surface
  • Very good sliding and dielectric characteristics
  • Oxygen value more than 95
  • Resistant to irradiation and weather
Perfect tubing for aggressive and pure liquids or gases. Can be sterilized in autoclaves.
Certificaten: Flame retardant according to UL94V0.