Tubing rolls, PFA, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S2030-12 S2030-14 S2030-18 S2030-24 S2030-28 S2030-30 S2032-02 S2032-04 S2032-05 S2032-06 S2032-10
BOHLS2030-12EA 223 EUR
BOHLS2030-12 BOHLS2030-14 BOHLS2030-18 BOHLS2030-24 BOHLS2030-28 BOHLS2030-30 BOHLS2032-02 BOHLS2032-04 BOHLS2032-05 BOHLS2032-06 BOHLS2032-10
Tubing rolls, PFA, BOLA
Slangen Transfer Tubing
Non-porous, transparent, gastight tubing.
  • Most universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Non-adhesive surface
  • Very good sliding and dielectric characteristics
  • Oxygen value more than 95
  • Resistant to irradiation and weather
  • Mechanical strength even at high temperatures
Perfect tubing for aggressive and pure liquids or gases. Can be sterilized in autoclaves.
Certificaten: Flame retardant according to UL94V0.