Multiple distributors with ground joint, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D623-14 D623-19 D623-45
BOHLD623-14EA 104 EUR
BOHLD623-14 BOHLD623-19 BOHLD623-45
Multiple distributors with ground joint, BOLA
Fittingen en connectoren
Three GL-threaded necks for connection of hard-walled tubing and tubes made of glass, plastic, or metal by means of BOLA laboratory screw joints.
  • Completely made of PTFE
  • Cone with elevated sealing rings on the outside
  • Knurled grip for easy removal
  • Universal chemical resistance
Easy extension of connections on glass lids with ground joint. For leading in and removal of liquids in glass reactors without contamination of the media through the ambient air. The product is only exposed to PTFE.