Rigid threaded couplings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H903-18 H903-14 H903-25
BOHLH903-18EA 173 EUR
BOHLH903-18 BOHLH903-14 BOHLH903-25
Rigid threaded couplings, BOLA
Fittingen en connectoren
Special design allows turning the two halfs of the coupling independently from each other, and to adjust connected components. This design also provides high stability. A sealing lip on the connecting tube with integrated FKM O-ring ensures good sealing on the GL thread.
  • Sturdy coupling with female GL threads made of PPS
  • Internal connecting tube made of PTFE
  • Universal chemical resistance
  • Flowing product is only exposed to PTFE
For connecting two GL threads of the same size.