VWR®, Polystyrene Assay Plates, 96- and 384-Well
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
732-3720EA 185 EUR
732-3720 732-3721 732-3722 732-3723 732-3724 732-3725 732-3728 732-3729 732-3730 732-3731 732-3734 732-3735 732-3736 732-3737 732-3738 732-3739 732-3740 732-3741 732-3742 732-3743 732-3744 732-3745 732-3746 732-3747 732-3748 732-3749 732-3750
VWR®, Polystyrene Assay Plates, 96- and 384-Well
Microplaten HTS-microplaten
Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
  • Flat bottom ideal for microscopic applications
  • V-bottom minimising residual liquid
  • U-bottom for optimising sample/cell collection
VWR® assay plates are available in clear, white or black polystyrene, with solid or clear bottom.
The bottom of the clear bottom plates is just 0,4 mm thick and extremely flat (±0,1 mm tolerance), ensuring high measurement precision and elimination of reading errors when performing cell-based assays using fluorescent or luminescent imaging. The tissue culture-treated plates are supplied lidded and sterile, in individual bags.
The bottom of the glass bottom plates is 175 µm thick, extremely flat with a tolerance of ±15 µm. They offer a higher performance than standard polystyrene plates for fluorescence assays, luminescence detection, scintillation counting and high resolution microscopy using confocal imaging.
The solid bottom plates are suitable for absorbance, fluorescence, luminescence and scintillation applications.
*Individual clear bottoms, used when light cross over from neighbouring wells needs to be minimised.