Built-in signalbox and switch amplifiers, SymLine®
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
108219 106590 106658 106678
SCTV108219EA 78.14 EUR
SCTV108219 SCTV106590 SCTV106658 SCTV106678
Built-in signalbox and switch amplifiers, SymLine®
Niveausensoren
Monitor two filling levels at once. Perfect integration into lab furniture with the Built-In signalbox and switch amplifier.
  • Monitor two filling levels at once
  • Touchpad control
  • Level control through status LEDs
  • Optical and acoustic alarm signals
  • Compatible with switch amplifier
LEDs and acoustic buzzer provide warning in due time before the filling level of collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via theMore Product Information