XF Matrix, CELLvo™
Leverancier: STEMBIOSYS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
XF-HPME-6WP XF-HPME-T75 XF-HPME-T150
STBSXF-HPME-6WPEA 0 EUR
STBSXF-HPME-6WP STBSXF-HPME-T75 STBSXF-HPME-T150
XF Matrix, CELLvo™
Celcultuursupplementen en -additieven
CELLvo™ XF Matrix is a ready to use cell culture substrate which provides a xeno-free native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.
  • Matrix made of polystyrene
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix proliferate more rapidly
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix maintain a smaller cell size
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ XF Matrix is a natural extracellular matrix synthesised in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow. CELLvo™ XF Matrix is a protein extracellular matrix produced under xeno-free conditions that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
Informatie over toebehoren: Accessories not included.
Leveringsinformatie: CELLvo™ XF Matrix arrives at room temperature but should be refrigerated at 2 to 8 °C upon arrival. Avoid extended exposure to light.
Verpakking: CELLvo™ XF Matrix is produced under aseptic conditions and should be handled using aseptic techniques.
Opgelet: CELLvo™ XF Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes. Handle CELLvo™ related products in accordance with established bio-safety practices.