Safety spectacles, Swiss One, Continental™
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1CONT21GR 1CONT23BB 1CONT23S
JSPA1CONT21GREA 11.3 EUR
JSPA1CONT21GR 111-2573 JSPA1CONT23S
Safety spectacles, Swiss One, Continental™
Brillen
Offering a wide field of vision and lightweight design, Continental™ safety spectacles feature a non-slip nose and endtips providing wearers with optimal comfort for all day wear.
  • Wraparound frame
  • Adjustable non-slip nose
  • Non-slip endtips
  • Lateral protection
Developed in five different versions, including gradient lenses, Blue blocker lenses, and AR lenses for an amazing light transmittance. Blue blocker lenses provide good protection when facing screens.
Certificaten: Compliant with: EN 166, EN 172. Frame: EN 166 FT CE. Lens: 5-1.7, 1 FT CE.