Laboratory glassware washers, SlimLine, PLW 6111
Leverancier: Miele
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
141-1517EA 24266 EUR
141-1516 141-1517 141-1518 141-1519 141-1534 141-1479 141-1513 141-1514 141-1515 141-1475 141-1497 141-1530 141-1476 141-1498 141-1531 141-1532 141-1477 141-1499 141-1533 141-1478 141-1482 141-1483 141-1484 141-1485 141-1480 141-1481 141-1527 141-1528 141-1529 141-1524 141-1525 141-1526 141-1486 141-1487 141-1488 141-1489 141-1493 141-1494 141-1495 141-1474 141-1496 141-1492
Laboratory glassware washers, SlimLine, PLW 6111
Wasmachines voor glaswerk
The PLW 6111 SlimLine laboratory washer offer a comprehensive and systematic solution to the reprocessing of laboratory glassware for analytical experiments.
  • Controls with flush, uninterrupted touch glass screen
  • All information visible at a glance thanks to 3,5" display
  • Maximum of 40 program slots (standard programs and vacant program slots)
  • Full glass door with integrated chamber lighting
  • Automatic door lock
  • Simple and effective cleaning
  • RS232 port for connecting to a printer or PC
The chamber, spray arms and tank filters are made from high grade stainless steel (DIN 1.4404/AISI 316L) with built-in softener*. Features heated boiler* for shorter cycle times and conductivity monitoring* for high level process security. It consists of HEPA H14 filter for sterile drying air.
Wide range of load carriers specifically designed for laboratory use. There is a maximum flexibility combined with intuitive use with a wide range of standard and configurable loading options. Possibility to reprocess even the smallest items of laboratory glassware through to very large containers (up to 50 L).
The innovative SmartLoad system enables chamber space to be used particularly efficiently. This is achieved by creating levels with different vertical clearances to perfectly match the goods requiring reprocessing.
The PLW 6111 series models are able to accommodate up to three baskets on telescopic racks. These baskets dock onto the water circulation system at four different heights. This allows a wide range of load height combinations, starting with using the lowest rack level only for a maximum vertical clearance of 630 mm.
Certificaten: Compliance with EN 61010-1, EN 601010-1, CSA approved.
Bestelinformatie: Baskets and accessories are not included, these must be ordered separately. Please contact your Avantor sales representative/technical services for assistance with the product configuration.
Opgelet: * depending on model