Nitrogen generator, Infinity 1051
Leverancier: Peak Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
564-0283EA 0 EUR
564-0283
Nitrogen generator, Infinity 1051
Gasgeneratoren Stikstofgeneratoren
Designed and engineered as an on-site, compressor less nitrogen generation solution tailored for the Shimadzu LCMS-8045, 8050 or 8060. The Infinity 1051 delivers one output of nitrogen and another of dry air to laboratories that already have an existing air supply.
  • Both a nitrogen and dry air output suitable for the Shimadzu LCMS-8045/50/60 and instruments requiring similar specifications
  • 24/7 operation at optimum performance if required
  • Gas is supplied on demand so generator works to your schedule
  • Few moving parts means little maintenance required and ensures long life of the generator
  • Minimal set-up required
  • Completely silent in operation
  • 1 year comprehensive on-site warranty