ESD safety sandals, BOA® Fit System, SCOTT BOA® Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
727651/38 727651/39 727651/40 727651/41 727651/42 727651/43 727651/44 727651/45 727651/46 727651/47 727651/48
ELTN727651/38EA 101.9 EUR
ELTN727651/38 ELTN727651/39 ELTN727651/40 ELTN727651/41 ELTN727651/42 ELTN727651/43 ELTN727651/44 ELTN727651/45 ELTN727651/46 ELTN727651/47 ELTN727651/48
ESD safety sandals, BOA® Fit System, SCOTT BOA® Low
Schoenen en laarzen
More volume in the ball, heel, and instep area makes this shoe particularly comfortable for people with wider feet.
  • Padded tongue
  • PU/PU tread sole SAFEGUARD
  • PU toe protection
  • Aluminium toe cap
  • BOA® Fit System closure
Microfibre/textile material with fabric lining for natural ventilation. Semi-orthopaedic inlay ESD sole in grey. Non-metallic, penetration-resistant midsole. Extra-wide version.
Certificaten: EN ISO 20345 S1P SRC, form A.