ESD safety shoes, lace-up, O1, LANA Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
974610/36 974610/37 974610/38 974610/39 974610/40 974610/41 974610/42
ELTN974610/36EA 88.9 EUR
ELTN974610/36 ELTN974610/37 ELTN974610/38 ELTN974610/39 ELTN974610/40 ELTN974610/41 ELTN974610/42
ESD safety shoes, lace-up, O1, LANA Low
Schoenen en laarzen
The shoe is ideally tailored to the ergonomics of female feet.
  • Padded tongue
  • Soft fleece ESD insole
  • TPU/PU sole MAXXIMO
  • Without protective toe cap
Microfibre/textile material with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD grey. Sole core made of Infinergy® by BASF.
Certificaten: EN ISO 20347 O1 FO/SRC, Form A.