ESD safety ankle boots, lace-up, jo_Clean Loop Mid, Jori
Leverancier: Elten
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Jori jo_Clean Loop
ELTN16651/39EA 57.9 EUR
ELTN16651/39 ELTN16651/38 ELTN16651/37 ELTN16651/36 ELTN16651/40 ELTN16641/47 ELTN16641/46 ELTN16641/48 ELTN16641/43 ELTN16641/42 ELTN16641/45 ELTN16641/44 ELTN16641/41 ELTN16641/40 ELTN16651/44 ELTN16651/43 ELTN16651/42 ELTN16651/41 ELTN16651/48 ELTN16651/47 ELTN16651/46 ELTN16651/45 ELTN16641/39 ELTN16641/36 ELTN16641/38 ELTN16641/37
ESD safety ankle boots, lace-up, jo_Clean Loop Mid, Jori
Schoenen en laarzen
Antistatic safety ankle boot made from hydrophobic microfiber and breathable textile lining and a full, padded bellows tongue.
  • MONO-PU sole jo_MONO
  • Composite toe cap
  • Metal- and leather-free
  • Available in black or white finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 SRC, form A.