ESD safety shoes, lace-up, S1P, jo_Speedy Low, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12121/36 12121/37 12121/38 12121/39 12121/40 12121/41 12121/42 12121/43 12121/44 12121/45 12121/46 12121/47 12121/48 12141/36 12141/37 12141/38 12141/39 12141/40 12141/41 12141/42 12141/43 12141/44 12141/45 12141/46 12141/47 12141/48 12131/36 12131/37 12131/38 12131/39 12131/40 12131/41 12131/42
ELTN12121/36EA 58.9 EUR
ELTN12121/36 ELTN12121/37 ELTN12121/38 ELTN12121/39 ELTN12121/40 ELTN12121/41 ELTN12121/42 ELTN12121/43 ELTN12121/44 ELTN12121/45 ELTN12121/46 ELTN12121/47 ELTN12121/48 ELTN12141/36 ELTN12141/37 ELTN12141/38 ELTN12141/39 ELTN12141/40 ELTN12141/41 ELTN12141/42 ELTN12141/43 ELTN12141/44 ELTN12141/45 ELTN12141/46 ELTN12141/47 ELTN12141/48 ELTN12131/36 ELTN12131/37 ELTN12131/38 ELTN12131/39 ELTN12131/40 ELTN12131/41 ELTN12131/42
ESD safety shoes, lace-up, S1P, jo_Speedy Low, Jori
Schoenen en laarzen
Excellent wearing comfort, and suitable for work in ESD sensitive or electrostatically protected areas.
  • Padded tongue
  • Non-metallic, penetration-resistant midsole
  • Rubber/EVA sole jo_Dynamic
  • Composite toe cap
Mesh material/microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Metal-free and leather-free.
Certificaten: EN ISO 20345 S1P HRO/SRC, form A.