ESD safety shoes, lace-up, S3, jo_Clean Low, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12641/36 12641/37 12641/38 12641/39 12641/40 12641/41 12641/42 12641/43 12641/44 12641/45 12641/46 12641/47 12641/48 12651/36 12651/37 12651/38 12651/39 12651/40 12651/41 12651/42 12651/43 12651/44 12651/45 12651/46 12651/47 12651/48
ELTN12641/36EA 55.9 EUR
ELTN12641/36 ELTN12641/37 ELTN12641/38 ELTN12641/39 ELTN12641/40 ELTN12641/41 ELTN12641/42 ELTN12641/43 ELTN12641/44 ELTN12641/45 ELTN12641/46 ELTN12641/47 ELTN12641/48 ELTN12651/36 ELTN12651/37 ELTN12651/38 ELTN12651/39 ELTN12651/40 ELTN12651/41 ELTN12651/42 ELTN12651/43 ELTN12651/44 ELTN12651/45 ELTN12651/46 ELTN12651/47 ELTN12651/48
ESD safety shoes, lace-up, S3, jo_Clean Low, Jori
Schoenen en laarzen
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • MONO-PU sole jo_MONO
  • Composite toe cap
  • Metal- and leather-free
  • Available in white or black finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 SRC, form A.