ESD safety ankle boots, pull-on, S3, jo_Clean Slipper Mid, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16671/36 16671/37 16671/38 16671/39 16671/40 16671/41 16671/42 16671/43 16671/44 16671/45 16671/46 16671/47 16671/48 16661/36 16661/37 16661/38 16661/39 16661/40 16661/41 16661/42 16661/43 16661/44 16661/45 16661/46 16661/47 16661/48
ELTN16671/36EA 57.9 EUR
ELTN16671/36 ELTN16671/37 ELTN16671/38 ELTN16671/39 ELTN16671/40 ELTN16671/41 ELTN16671/42 ELTN16671/43 ELTN16671/44 ELTN16671/45 ELTN16671/46 ELTN16671/47 ELTN16671/48 ELTN16661/36 ELTN16661/37 ELTN16661/38 ELTN16661/39 ELTN16661/40 ELTN16661/41 ELTN16661/42 ELTN16661/43 ELTN16661/44 ELTN16661/45 ELTN16661/46 ELTN16661/47 ELTN16661/48
ESD safety ankle boots, pull-on, S3, jo_Clean Slipper Mid, Jori
Schoenen en laarzen
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full bellows tongue
  • MONO-PU sole jo_MONO
  • Composite toe cap
  • Metal- and leather-free
  • Available in black or white finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 SRC, Form B.