Utility Boxes
Leverancier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1752 1753 1754 1755 3629 3630 3631
216-2472EA 39.5 EUR
216-2472 216-2473 216-2474 216-2475 216-2477 216-2478 216-2479
Utility Boxes
Dozen Opbergdozen
Range of practical PP boxes in 9 different sizes, 5 round and 4 square, ranging from 0,25 to 8 L.
  • Food grade
  • Soft and flexible lids
  • Anti-slip surface suitable for freezer and fridge
  • Acid and alkali resistant
  • Cold and heat resistant: –20 to +100 °C
All the boxes have a soft and flexible lid, ensuring a fastened closure as well as easy assembling and removal.
The surface of the boxes is slightly roughed and offers a 'anti-slip' function even with wet hands.
Easy to clean, these utility laboratory storage boxes are stackable and can be placed inMore Product Information