ESD safety ankle boots, lace-up, jo_Rapid Mid, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16021/36 16021/37 16021/38 16021/39 16021/40 16021/41 16021/42 16021/43 16021/44 16021/45 16021/46 16021/47 16021/48
ELTN16021/36EA 66.9 EUR
ELTN16021/36 ELTN16021/37 ELTN16021/38 ELTN16021/39 ELTN16021/40 ELTN16021/41 ELTN16021/42 ELTN16021/43 ELTN16021/44 ELTN16021/45 ELTN16021/46 ELTN16021/47 ELTN16021/48
ESD safety ankle boots, lace-up, jo_Rapid Mid, Jori
Schoenen en laarzen
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/PU sole jo_ATHLETIC
  • Rubber scuff cap
  • Composite toe cap
  • Metal- and leather-free
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 HRO/SRC, form B.