ESD safety shoes, elastic lacing system, Impulse Lady XXTL Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
742301/35 742301/36 742301/37 742301/38 742301/39 742301/40 742301/41 742301/42
ELTN742301/35EA 88.9 EUR
ELTN742301/35 ELTN742301/36 ELTN742301/37 ELTN742301/38 ELTN742301/39 ELTN742301/40 ELTN742301/41 ELTN742301/42
ESD safety shoes, elastic lacing system, Impulse Lady XXTL Low
Schoenen en laarzen
Ladies' safety shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • PU/PU sole WELLMAXX TRAINERS LADY
  • Steel toe cap
  • Elastic lacing system with individual quick locking system
  • Leather-free
Microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole, PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certificaten: EN ISO 20345 S1P SRC, form A.