ESD safety shoes, lace-up, S1P, Broker XXB Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
729311/39 729311/40 729311/41 729311/42 729311/43 729311/44 729311/45 729311/46 729311/47 729311/48 729311/49
ELTN729311/39EA 99.9 EUR
ELTN729311/39 ELTN729311/40 ELTN729311/41 ELTN729311/42 ELTN729311/43 ELTN729311/44 ELTN729311/45 ELTN729311/46 ELTN729311/47 ELTN729311/48 ELTN729311/49
ESD safety shoes, lace-up, S1P, Broker XXB Low
Schoenen en laarzen
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • MONO-PU sole WELLMAXX BUSINESS
  • Steel toe cap
  • Leather-free
Microfibre construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certificaten: EN ISO 20345 S1P SRC, form A.