ESD safety shoes, lace-up, jo_Swift Low, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12051/36 12051/37 12051/38 12051/39 12051/40 12051/41 12051/42 12051/43 12051/44 12051/45 12051/46 12051/47 12051/48 12061/36 12061/37 12061/38 12061/39 12061/40 12061/41 12061/42 12061/43 12061/44 12061/45 12061/46 12061/47 12061/48 12071/36 12071/37 12071/38 12071/39 12071/40 12071/41 12071/42 12071/43 12071/44 12071/45 12071/46 12071/47 12071/48 12091/36 12091/37 12091/38 12091/39 12091/40 12091/41 12091/42 12091/43 12091/44 12091/45 12091/46 12091/47 12091/48
ELTN12051/36EA 63.9 EUR
ELTN12051/36 ELTN12051/37 ELTN12051/38 ELTN12051/39 ELTN12051/40 ELTN12051/41 ELTN12051/42 ELTN12051/43 ELTN12051/44 ELTN12051/45 ELTN12051/46 ELTN12051/47 ELTN12051/48 ELTN12061/36 ELTN12061/37 ELTN12061/38 ELTN12061/39 ELTN12061/40 ELTN12061/41 ELTN12061/42 ELTN12061/43 ELTN12061/44 ELTN12061/45 ELTN12061/46 ELTN12061/47 ELTN12061/48 ELTN12071/36 ELTN12071/37 ELTN12071/38 ELTN12071/39 ELTN12071/40 ELTN12071/41 ELTN12071/42 ELTN12071/43 ELTN12071/44 ELTN12071/45 ELTN12071/46 ELTN12071/47 ELTN12071/48 ELTN12091/36 ELTN12091/37 ELTN12091/38 ELTN12091/39 ELTN12091/40 ELTN12091/41 ELTN12091/42 ELTN12091/43 ELTN12091/44 ELTN12091/45 ELTN12091/46 ELTN12091/47 ELTN12091/48
ESD safety shoes, lace-up, jo_Swift Low, Jori
Schoenen en laarzen
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/PU sole jo_ATHLETIC
  • Composite toe cap
  • Metal- and leather-free
  • Available in multiple colour options
Textile material construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S1P HRO/SRC, form A.