ESD safety ankle boots, BOA® Fit System, HANNAH XXTL BOA® Mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
746561/35 746561/36 746561/37 746561/38 746561/39 746561/40 746561/41 746561/42
ELTN746561/35EA 103.9 EUR
ELTN746561/35 ELTN746561/36 ELTN746561/37 ELTN746561/38 ELTN746561/39 ELTN746561/40 ELTN746561/41 ELTN746561/42
ESD safety ankle boots, BOA® Fit System, HANNAH XXTL BOA® Mid
Schoenen en laarzen
Ladies' safety ankle boot is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • PU/PU sole, WELLMAXX TRAINERS LADY
  • Steel toe cap
  • BOA® Fit System
  • Leather-free
Hydrophobised microfibre/hydrophobised textile material construction with fabric lining for natural ventilation. Full-length inlay sole, ESD PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certificaten: EN ISO 20345 S3 SRC, form B.