Chemical-resistant labels for resin embedding
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
XST-251C1-2WH XST-251C3-10WH
GAINXST-251C1-2WHEA 96.2 EUR
GAINXST-251C1-2WH GAINXST-251C3-10WH
Chemical-resistant labels for resin embedding
Etiketten
Chemical-resistant, thermal-transfer labels for resin embedding, commonly used for transmission electron microscopy (TEM).
  • Labels and printout withstand resin infiltration and heat polymerization up to 150 °C (302 °F)
  • Small size accommodates various sizes of embedding capsules and molds
  • Will withstand alcohol, xylene, toluene, acetone, DMSO, as well as other harsh organic solvents
  • Available on 2,5 or 7,6 cm cores for printing with all common thermal-transfer printer brands
The novel design, and small size of the ResiTAG™ label can accommodate variousMore Product Information
Order Now
Specificaties
Environmentally Preferable