Transparent cryogenic labels for thermal-transfer printers
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HBCL-18BMC1-2 HBCL-18BMC3-2
GAINHBCL-18BMC1-2EA 90.1 EUR
GAINHBCL-18BMC1-2 GAINHBCL-18BMC3-2
Transparent cryogenic labels for thermal-transfer printers
Etiketten
Transparent thermoplastic cryogenic labels for bio-medical research and clinical laboratories.
  • Labels can withstand a wide temperature range: extreme low (–196 °C) to elevated (100 °C)
  • Transparent design ensures contents of labeled containers remain visible
  • Printable with all major thermal-transfer printer brands
Labels will withstand long-term storage in deep-freeze and harsh laboratory environments, including liquid and vapor phase liquid nitrogen at –196 °C (–320.8 °F), –80 °C (–112 °F) freezers, and dry ice transportation.