Cryogenic Lamination Tapes
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLAM-19C3-200 CLAM-25C3-200 CLAM-50C3-200
GAINCLAM-19C3-200EA 48.1 EUR
GAINCLAM-19C3-200 GAINCLAM-25C3-200 GAINCLAM-50C3-200
Cryogenic Lamination Tapes
Tape
Thin cryo lamination tape provides added protection against abrasion, harsh chemicals, and extreme temperatures. Designed to withstand long-term storage in liquid nitrogen and ultra-low temperature freezers, multiple freeze-thaw cycles, and transport on dry ice. This clear tape can be applied over existing labels.
  • Temperature range: −196 to +93 °C
  • Clear lamination tape protects material from extreme cryogenic conditions, including liquid phase and vapour phase liquid nitrogen
  • Resists harsh laboratory environments, including multiple freeze-thaw cycles
More Product Information
Order Now
Specificaties
Environmentally Preferable