Elbow connector for waste pipes, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1180-01 R1182-01
BOHLR1180-01EA 143 EUR
BOHLR1180-01 BOHLR1182-01
Elbow connector for waste pipes, b.safe
Fittingen en connectoren
Elbow tube approx. 90° made of static dissipative PE-EX.
Connection 1: Lock nut and clamp ring for inserting and clamping b.safe Waste Pipes with Ext.Ø 32 mm.
Connection 2: Either lock nut and clamp ring for clamping b.safe Waste Pipes or male thread GL 25 for transition to flexible b.safe Waste Tubes.